CAS RN 261951-85-3
IUPAC Name:
1-bromo-3-fluoro-2-(trifluoromethyl)benzene
Synonyms:
1-bromo-3-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol):
243
Molecular Formula:
C7H3BrF4
InChi Key:
LOXSSZUHCPBDMD-UHFFFAOYSA-N
SMILES:
FC1=C(C(Br)=CC=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Bromo-6-fluorobenzotrifluoride 97.0+%, TCI America™
CAS: 261951-85-3 Molecular Formula: C7H3BrF4 Molecular Weight (g/mol): 242.999 MDL Number: MFCD01631569 InChI Key: LOXSSZUHCPBDMD-UHFFFAOYSA-N Synonym: 2-bromo-6-fluorobenzotrifluoride,1-bromo-3-fluoro-2-trifluoromethyl benzene,3-fluoro-2-trifluoromethyl bromobenzene,benzene, 1-bromo-3-fluoro-2-trifluoromethyl,2-bromo-alpha,alpha,alpha,6-tetrafluorotoluene,1-bromo-3-fluoro-2-trifluoromethyl-benzene,pubchem1638,2-bromo-6-fluorobenzo trifluoride PubChem CID: 2773345 IUPAC Name: 1-bromo-3-fluoro-2-(trifluoromethyl)benzene SMILES: C1=CC(=C(C(=C1)Br)C(F)(F)F)F