missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzotrifluoride 97.0+%, TCI America™

Catalog No. B48331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48331G 1 g
B48335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48331G Supplier TCI America Supplier No. B48331G
Only null left

Chemical Identifiers

CAS 261951-85-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD01631569
InChI Key LOXSSZUHCPBDMD-UHFFFAOYSA-N
Synonym 2-bromo-6-fluorobenzotrifluoride, 1-bromo-3-fluoro-2-trifluoromethyl benzene, 3-fluoro-2-trifluoromethyl bromobenzene, benzene, 1-bromo-3-fluoro-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha,6-tetrafluorotoluene, 1-bromo-3-fluoro-2-trifluoromethyl-benzene, pubchem1638, 2-bromo-6-fluorobenzo trifluoride
PubChem CID 2773345
IUPAC Name 1-bromo-3-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)Br)C(F)(F)F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 243.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-fluorobenzotrifluoride