missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. B40175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40175G 5 g
B40171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40175G Supplier TCI America Supplier No. B40175G
Only null left

Chemical Identifiers

CAS 2252-37-1
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD01569539
InChI Key MDAZJVAIZVUWDE-UHFFFAOYSA-N
Synonym 2-fluoro-6-bromobenzoic acid, 6-bromo-2-fluorobenzoic acid, benzoic acid, 2-bromo-6-fluoro, 2-bromo-6-fluorobenzoicacid, 2-bromo-6-fluoro-benzoic acid, buttpark 24\01-96, pubchem1317, 2-bromo-6-fluorobenzoic, intermediates-zcf02603, acmc-209fx2
PubChem CID 302621
IUPAC Name 2-bromo-6-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)C(=O)O)F

Specifications

Melting Point 156°C
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-fluorobenzoic Acid