accessibility menu, dialog, popup

UserName

HOOFBr

CAS RN 2252-37-1

IUPAC Name: 2-bromo-6-fluorobenzoic acid
Synonyms: 2-bromo-6-fluorobenzoic acid
Molecular Weight (g/mol): 219.01
Molecular Formula: C7H4BrFO2
InChi Key: MDAZJVAIZVUWDE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC=C1Br
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)