missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog No. p-7035354
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5307103 1 g
AAH5307106 5 g
AAH5307114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5307103 Supplier Thermo Scientific Chemicals Supplier No. H5307103
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 913835-80-0
Molecular Formula C6H5BBrFO2
Molecular Weight (g/mol) 218.816
MDL Number MFCD05664309
InChI Key MVSHYHSMIRBRGU-UHFFFAOYSA-N
Synonym 2-bromo-6-fluorophenyl boronic acid, 2-bromo-6-fluorobenzeneboronic acid, 2-bromo-6-fluorophenylboronicacid, boronic acid, b-2-bromo-6-fluorophenyl, acmc-209ra3, 6-bromo-2-fluorophenylboronic acid, 2-bromanyl-6-fluoranyl-phenyl boronic acid
PubChem CID 16217456
IUPAC Name (2-bromo-6-fluorophenyl)boronic acid
SMILES B(C1=C(C=CC=C1Br)F)(O)O

Specifications

Melting Point 142°C to 145°C
Quantity 1 g
Formula Weight 218.82
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures