accessibility menu, dialog, popup

UserName

BBrFHOOH

CAS RN 913835-80-0

IUPAC Name: (2-bromo-6-fluorophenyl)boronic acid
Synonyms: 2-bromo-6-fluorophenylboronic acid
Molecular Weight (g/mol): 218.82
Molecular Formula: C6H5BBrFO2
InChi Key: MVSHYHSMIRBRGU-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results