missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)phenol, 98%

Catalog No. AAH6400914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6400914 25 g
AAH6400906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6400914 Supplier Thermo Scientific Chemicals Supplier No. H6400914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 402-05-1 | C7H4BrF3O | 241.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-05-1
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD09261190
InChI Key INHVNZLKNPJCJD-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl phenol, phenol, 2-bromo-5-trifluoromethyl, acmc-209jbu, phenol,2-bromo-5-trifluoromethyl, 3-bromo-5-hydroxybenzotrifluoride, 4-bromo-3-hydroxybenzotrifluoride
PubChem CID 21914470
IUPAC Name 2-bromo-5-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)O)Br

Specifications

Color Brown
Quantity 25 g
UN Number UN3265
Formula Weight 241.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)phenol
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only