accessibility menu, dialog, popup

UserName

FFFHOBr

CAS RN 402-05-1

IUPAC Name: 2-bromo-5-(trifluoromethyl)phenol
Synonyms: 2-bromo-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: INHVNZLKNPJCJD-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results