missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™

Catalog No. b45085g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45085G 5 g
B45081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45085G Supplier TCI America Supplier No. B45085G
Only null left

Chemical Identifiers

CAS 102684-91-3
Molecular Formula C8H4BrF3O
Molecular Weight (g/mol) 253.02
MDL Number MFCD04973760
InChI Key CSOBJYGHQOLWOD-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl benzaldehyde, 4-bromo-3-formylbenzotrifluoride, 2-bromo-5-1,1-dimethylethyl-benzaldehyde, benzaldehyde, 2-bromo-5-1,1-dimethylethyl, benzaldehyde, 2-bromo-5-trifluoromethyl, paragos 530289, pubchem1428, pubchem4179, ksc498c1d, bromo 5-trifluoromethyl benzaldehyde
PubChem CID 7018047
IUPAC Name 2-bromo-5-(trifluoromethyl)benzaldehyde
SMILES FC(F)(F)C1=CC(C=O)=C(Br)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 253.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)benzaldehyde
TSCA No
Recommended Storage Refrigerator