accessibility menu, dialog, popup

UserName

CAS RN 102684-91-3

IUPAC Name: 2-bromo-5-(trifluoromethyl)benzaldehyde
Synonyms: 2-bromo-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 253.02
Molecular Formula: C8H4BrF3O
InChi Key: CSOBJYGHQOLWOD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(C=O)=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)