missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethoxy)phenol 98.0+%, TCI America™

Catalog No. p-7119956
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31085G 5 g
B310825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31085G Supplier TCI America Supplier No. B31085G

Chemical Identifiers

CAS 205371-26-2
Molecular Formula C7H4BrF3O2
Molecular Weight (g/mol) 257.01
MDL Number MFCD06660162
InChI Key RHRRKORKKIVAGJ-UHFFFAOYSA-N
PubChem CID 17750742
IUPAC Name 2-bromo-5-(trifluoromethoxy)phenol
SMILES OC1=C(Br)C=CC(OC(F)(F)F)=C1

Specifications

Color Yellow
Quantity 5 g
Synonym 2-bromo-5-trifluoromethoxy phenol, phenol, 2-bromo-5-trifluoromethoxy, 2-hydroxy-4-trifluoromethoxy bromobenzene, 1-bromo-2-hydroxy-4-trifluoromethoxy benzene, pubchem15610, acmc-209fbt, rhrrkorkkivagj-uhfffaoysa, phenol,2-bromo-5-trifluoromethoxy, 2-bromanyl-5-trifluoromethyloxy phenol
Formula Weight 257.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-(trifluoromethoxy)phenol