accessibility menu, dialog, popup

UserName

OFFFHOBr

CAS RN 205371-26-2

IUPAC Name: 2-bromo-5-(trifluoromethoxy)phenol
Synonyms: 2-bromo-5-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 257.01
Molecular Formula: C7H4BrF3O2
InChi Key: RHRRKORKKIVAGJ-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=CC(OC(F)(F)F)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)