missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethoxy)phenol, 97%

Catalog No. AAH2638906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2638906 5 g
AAH2638914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2638906 Supplier Thermo Scientific Chemicals Supplier No. H2638906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 205371-26-2 | C7H4BrF3O2 | 257.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 205371-26-2
Molecular Formula C7H4BrF3O2
Molecular Weight (g/mol) 257.01
MDL Number MFCD06660162
InChI Key RHRRKORKKIVAGJ-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethoxy phenol, phenol, 2-bromo-5-trifluoromethoxy, 2-hydroxy-4-trifluoromethoxy bromobenzene, 1-bromo-2-hydroxy-4-trifluoromethoxy benzene, pubchem15610, acmc-209fbt, rhrrkorkkivagj-uhfffaoysa, phenol,2-bromo-5-trifluoromethoxy, 2-bromanyl-5-trifluoromethyloxy phenol
PubChem CID 17750742
IUPAC Name 2-bromo-5-(trifluoromethoxy)phenol
SMILES OC1=C(Br)C=CC(OC(F)(F)F)=C1

Specifications

Refractive Index 1.4815
Quantity 5 g
Formula Weight 257.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethoxy)phenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only