missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™

Catalog No. p-7124155
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5674200MG 200 mg
B56741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5674200MG Supplier TCI America Supplier No. B5674200MG
Only null left

Chemical Identifiers

CAS 10368-73-7
Molecular Formula C24H17Br
Molecular Weight (g/mol) 385.30
MDL Number MFCD00092320
InChI Key JQAGNDRJKSDTEU-UHFFFAOYSA-N
Synonym 2-Bromo-1,3,5-triphenylbenzene, 2′-Bromo-5′-phenyl-m-terphenyl
PubChem CID 292152
IUPAC Name 2-bromo-3,5-diphenyl-1,1'-biphenyl
SMILES BrC1=C(C=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 130°C
Color White
Quantity 200 mg
Formula Weight 385.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Bromo-5′-phenyl-1,1′:3′,1′′-terphenyl