CAS RN 10368-73-7
IUPAC Name:
2-bromo-3,5-diphenyl-1,1'-biphenyl
Synonyms:
2-bromo-3,5-diphenyl-1,1'-biphenyl
Molecular Weight (g/mol):
385.3
Molecular Formula:
C24H17Br
InChi Key:
JQAGNDRJKSDTEU-UHFFFAOYSA-N
SMILES:
BrC1=C(C=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2'-Bromo-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™
CAS: 10368-73-7 Molecular Formula: C24H17Br Molecular Weight (g/mol): 385.30 MDL Number: MFCD00092320 InChI Key: JQAGNDRJKSDTEU-UHFFFAOYSA-N Synonym: 2-Bromo-1,3,5-triphenylbenzene, 2′-Bromo-5′-phenyl-m-terphenyl PubChem CID: 292152 IUPAC Name: 2-bromo-3,5-diphenyl-1,1'-biphenyl SMILES: BrC1=C(C=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1