missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123195
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38415G 5 g
B384125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38415G Supplier TCI America Supplier No. B38415G
Only null left

Chemical Identifiers

CAS 943-14-6
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00134558
InChI Key UVFWYVCDRKRAJH-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179
PubChem CID 243025
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrobenzoic Acid