accessibility menu, dialog, popup

UserName

HOOBrNOO

CAS RN 943-14-6

IUPAC Name: 2-bromo-5-nitrobenzoic acid
Synonyms: 2-bromo-5-nitrobenzoic acid
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: UVFWYVCDRKRAJH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1Br)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)