missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitroaniline 97.0+%, TCI America™

Catalog No. b304125g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B304125G 25 g
B30415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B304125G Supplier TCI America Supplier No. B304125G
Only null left

Chemical Identifiers

CAS 10403-47-1
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD00051578
InChI Key BAAUCXCLMDAZEL-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-nitro, 2-bromo-5-nitrobenzenamine, 3-amino-4-bromonitrobenzene, 2-bromo-5-nitro-phenylamine, 2-bromo-5-nitrophenylamine, pubchem21903, 2-bromo 5-nitroaniline, 2-bromo-5-nitro-aniline, 2-bromo-5-nitro aniline, acmc-2098ai
PubChem CID 82607
IUPAC Name 2-bromo-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)Br

Specifications

Melting Point 140°C
Quantity 25 g
Formula Weight 217.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitroaniline