accessibility menu, dialog, popup

UserName

NH2BrNOO

CAS RN 10403-47-1

IUPAC Name: 2-bromo-5-nitroaniline
Synonyms: benzenamine, 2-bromo-5-nitro-
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: BAAUCXCLMDAZEL-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Br)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)