missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorophenyl acetic acid, 96%, Thermo Scientific Chemicals

Catalog No. p-7054098
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6200506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6200506 Supplier Thermo Scientific Chemicals Supplier No. H6200506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 739336-26-6 | Molecular Formula: C8H6BrFO2 | Molecular Weight: 233.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 739336-26-6
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD03094440
InChI Key BPNWLZNAKZSBHH-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorophenylacetic acid, 2-2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenylaceticacid, 2-bromo-5-fulorophenylacetic acid, benzeneacetic acid, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenyl-acetic acid, pubchem16327, 2-2-bromo-5-fluoro-phenyl acetic acid, acmc-209osx
PubChem CID 2773927
IUPAC Name 2-(2-bromo-5-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1F)CC(=O)O)Br

Specifications

Melting Point 113°C to 114°C
Quantity 5 g
Formula Weight 233.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-fluorophenyl acetic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only