accessibility menu, dialog, popup

UserName

CAS RN 739336-26-6

IUPAC Name: 2-(2-bromo-5-fluorophenyl)acetic acid
Synonyms: (2-bromo-5-fluorophenyl)acetic acid
Molecular Weight (g/mol): 233.04
Molecular Formula: C8H6BrFO2
InChi Key: BPNWLZNAKZSBHH-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(Br)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results