missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. B37845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37845G 5 g
B378425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37845G Supplier TCI America Supplier No. B37845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112399-50-5
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD00042187
InChI Key CZLWYKAZAVYQIK-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzyl bromide, 1-bromo-2-bromomethyl-4-fluorobenzene, 5-fluoro-2-bromobenzyl bromide, 2-bromo-5-fluorobenzylbromide, benzene, 1-bromo-2-bromomethyl-4-fluoro, alpha,2-dibromo-5-fluorotoluene, pubchem1953, acmc-1bt22, ksc493i2h, 2-bromo-5-fluoro benzyl bromide
PubChem CID 517972
IUPAC Name 1-bromo-2-(bromomethyl)-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)CBr)Br

Specifications

Color White
Boiling Point 96°C
Quantity 5 g
UN Number 3261
Formula Weight 267.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-5-fluorobenzyl Bromide