accessibility menu, dialog, popup

UserName

FBrBr

CAS RN 112399-50-5

IUPAC Name: 1-bromo-2-(bromomethyl)-4-fluorobenzene
Synonyms: 1-bromo-2-(bromomethyl)-4-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: CZLWYKAZAVYQIK-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)