missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzyl bromide, 97%

Catalog No. AAL0955303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0955303 1 g
AAL0955306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0955303 Supplier Thermo Scientific Chemicals Supplier No. L0955303

CAS: 112399-50-5 | C7H5Br2F | 267.923 g/mol

It is an important pharmaceutical intermediate. Alkylation of the β-amino ester 1a with 2-bromo-5-fluorobenzyl bromide yields Benzazepines as a white solid after silica gel chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate. Alkylation of the β-amino ester 1a with 2-bromo-5-fluorobenzyl bromide yields Benzazepines as a white solid after silica gel chromatography.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.

Chemical Identifiers

CAS 112399-50-5
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD00042187
InChI Key CZLWYKAZAVYQIK-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzyl bromide, 1-bromo-2-bromomethyl-4-fluorobenzene, 5-fluoro-2-bromobenzyl bromide, 2-bromo-5-fluorobenzylbromide, benzene, 1-bromo-2-bromomethyl-4-fluoro, alpha,2-dibromo-5-fluorotoluene, pubchem1953, acmc-1bt22, ksc493i2h, 2-bromo-5-fluoro benzyl bromide
PubChem CID 517972
IUPAC Name 1-bromo-2-(bromomethyl)-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)CBr)Br

Specifications

Melting Point 33°C to 36°C
Boiling Point 89°C to 90°C (1 mmHg)
Refractive Index 1.59
Quantity 1 g
UN Number UN3265
Beilstein 4177658
Solubility Information Difficult to mix in water.
Formula Weight 267.92
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluorobenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only