missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluorobenzoic Acid 98.0+%, TCI America™
$180.97 - $592.95
Chemical Identifiers
| CAS | 394-28-5 |
|---|---|
| Molecular Formula | C7H3BrFO2 |
| Molecular Weight (g/mol) | 218.00 |
| MDL Number | MFCD00142874 |
| InChI Key | OQBMJMJZMDBQSM-UHFFFAOYSA-M |
| Synonym | benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 |
| PubChem CID | 2778181 |
| IUPAC Name | 2-bromo-5-fluorobenzoate |
| SMILES | [O-]C(=O)C1=CC(F)=CC=C1Br |
Chemical Identifiers
| 394-28-5 | |
| 218.00 | |
| OQBMJMJZMDBQSM-UHFFFAOYSA-M | |
| 2778181 | |
| [O-]C(=O)C1=CC(F)=CC=C1Br |
| C7H3BrFO2 | |
| MFCD00142874 | |
| benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 | |
| 2-bromo-5-fluorobenzoate |
Specifications
| 394-28-5 | |
| White-Yellow | |
| MFCD00142874 | |
| benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 | |
| [O-]C(=O)C1=CC(F)=CC=C1Br | |
| 218.00 | |
| 219.01 | |
| Crystalline Powder |
| 156°C | |
| C7H3BrFO2 | |
| 5 g | |
| OQBMJMJZMDBQSM-UHFFFAOYSA-M | |
| 2-bromo-5-fluorobenzoate | |
| 2778181 | |
| ≥98.0% (GC,T) | |
| 2-Bromo-5-fluorobenzoic Acid |
Safety and Handling
TSCA : No