accessibility menu, dialog, popup

UserName

OOFBr

CAS RN 394-28-5

IUPAC Name: 2-bromo-5-fluorobenzoate
Synonyms: 2-bromo-5-fluorobenzoate
Molecular Weight (g/mol): 218
Molecular Formula: C7H3BrFO2
InChi Key: OQBMJMJZMDBQSM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=CC=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)