missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. B27225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27225G 5 g
B272225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27225G Supplier TCI America Supplier No. B27225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 394-28-5
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00142874
InChI Key OQBMJMJZMDBQSM-UHFFFAOYSA-M
Synonym benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
PubChem CID 2778181
IUPAC Name 2-bromo-5-fluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=CC=C1Br

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-fluorobenzoic Acid