missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluorobenzoic acid, 98+%
CAS: 394-28-5 | C7H3BrFO2 | 218.00 g/mol
$225.96 - $958.56
Chemical Identifiers
| CAS | 394-28-5 |
|---|---|
| Molecular Formula | C7H3BrFO2 |
| Molecular Weight (g/mol) | 218.00 |
| MDL Number | MFCD00142874 |
| InChI Key | OQBMJMJZMDBQSM-UHFFFAOYSA-M |
| Synonym | benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 |
| PubChem CID | 2778181 |
| IUPAC Name | 2-bromo-5-fluorobenzoic acid |
| SMILES | [O-]C(=O)C1=CC(F)=CC=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2539406
|
Thermo Scientific Chemicals
B2539406 |
5 g |
Each for $225.96
|
|
|||||
|
AAB2539414
|
Thermo Scientific Chemicals
B2539414 |
25 g |
Each for $958.56
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 394-28-5 | |
| 218.00 | |
| OQBMJMJZMDBQSM-UHFFFAOYSA-M | |
| 2778181 | |
| [O-]C(=O)C1=CC(F)=CC=C1Br |
| C7H3BrFO2 | |
| MFCD00142874 | |
| benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 | |
| 2-bromo-5-fluorobenzoic acid |
Specifications
| 394-28-5 | |
| C7H3BrFO2 | |
| 5 g | |
| benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747 | |
| [O-]C(=O)C1=CC(F)=CC=C1Br | |
| 218.00 | |
| 219.01 | |
| 2-Bromo-5-fluorobenzoic acid |
| 154°C to 158°C | |
| MFCD00142874 | |
| 2575978 | |
| OQBMJMJZMDBQSM-UHFFFAOYSA-M | |
| 2-bromo-5-fluorobenzoic acid | |
| 2778181 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : Yes
RUO – Research Use Only