missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-5'-fluoroacetophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6126103

 View more versions of this product

Catalog No. AAH6126103

Only null left
Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Bromo-5′-fluoroacetophenone is used in the synthesis of oxabicyclic compounds. It is also used in the enantioselective synthesis of chiral 3-aryl-1-indanone molecules.

Solubility
Soluble in methanol.

Notes
Light Sensitive. Store in cool, well ventilated area. Keep container tightly closed.
Specifications

Specifications

2'-Bromo-5'-fluoroacetophenone
144°C (18 mmHg)
1.5525
1 g
Soluble in methanol.
CC(=O)C1=C(Br)C=CC(F)=C1
217.04
217.04
1006-33-3
C8H6BrFO
MFCD07780635
2'-bromo-5'-fluoroacetophenone, 1-2-bromo-5-fluorophenyl ethanone, 2-bromo-5-fluoroacetophenone, 1-2-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-5-fluorophenyl, 1-2-bromo-5-fluoro-phenyl ethanone, ethanone,1-2-bromo-5-fluorophenyl, pubchem3214, acmc-1btyq, ksc499c4f
BQXROZQUDCJBBY-UHFFFAOYSA-N
1-(2-bromo-5-fluorophenyl)ethanone
22736080
98%

Chemical Identifiers

1006-33-3
217.04
BQXROZQUDCJBBY-UHFFFAOYSA-N
1-(2-bromo-5-fluorophenyl)ethanone
C8H6BrFO
MFCD07780635
22736080
CC(=O)C1=C(Br)C=CC(F)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions