missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-5'-fluoroacetophenone, 98%, Thermo Scientific™

Catalog No. p-7041189
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61261MD 250 mg
AAH6126103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61261MD Supplier Thermo Scientific Chemicals Supplier No. H61261MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Bromo-5′-fluoroacetophenone is used in the synthesis of oxabicyclic compounds. It is also used in the enantioselective synthesis of chiral 3-aryl-1-indanone molecules.

Solubility
Soluble in methanol.

Notes
Light Sensitive. Store in cool, well ventilated area. Keep container tightly closed.

Chemical Identifiers

CAS 1006-33-3
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD07780635
InChI Key BQXROZQUDCJBBY-UHFFFAOYSA-N
PubChem CID 22736080
IUPAC Name 1-(2-bromo-5-fluorophenyl)ethanone
SMILES CC(=O)C1=C(Br)C=CC(F)=C1

Specifications

Boiling Point 144°C (18 mmHg)
Refractive Index 1.5525
Quantity 250 mg
Synonym 2'-bromo-5'-fluoroacetophenone, 1-2-bromo-5-fluorophenyl ethanone, 2-bromo-5-fluoroacetophenone, 1-2-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-5-fluorophenyl, 1-2-bromo-5-fluoro-phenyl ethanone, ethanone,1-2-bromo-5-fluorophenyl, pubchem3214, acmc-1btyq, ksc499c4f
Solubility Information Soluble in methanol.
Formula Weight 217.04
Percent Purity 98%
Chemical Name or Material 2'-Bromo-5'-fluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures