CAS RN 1006-33-3
IUPAC Name:
1-(2-bromo-5-fluorophenyl)ethan-1-one
Synonyms:
1-(2-bromo-5-fluorophenyl)ethanone
Molecular Weight (g/mol):
217.04
Molecular Formula:
C8H6BrFO
InChi Key:
BQXROZQUDCJBBY-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(Br)C=CC(F)=C1
1
–
1
of
1
results
1
Promotions Available
2'-Bromo-5'-fluoroacetophenone, 98%, Thermo Scientific™
2'-Bromo-5'-fluoroacetophenone, 98%, Thermo Scientific™
CAS: 1006-33-3 Molecular Formula: C8H6BrFO Molecular Weight (g/mol): 217.04 MDL Number: MFCD07780635 InChI Key: BQXROZQUDCJBBY-UHFFFAOYSA-N PubChem CID: 22736080 IUPAC Name: 1-(2-bromo-5-fluorophenyl)ethanone SMILES: CC(=O)C1=C(Br)C=CC(F)=C1