missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorobenzaldehyde 97.0+%, TCI America™

Catalog No. B29555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29555G 5 g
B295525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29555G Supplier TCI America Supplier No. B29555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174265-12-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
InChI Key IIISHLMCTDMUHH-UHFFFAOYSA-N
Synonym 5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
PubChem CID 15391279
IUPAC Name 2-bromo-5-chlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)Br

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-chlorobenzaldehyde