accessibility menu, dialog, popup

UserName

ClOBr

CAS RN 174265-12-4

IUPAC Name: 2-bromo-5-chlorobenzaldehyde
Synonyms: 2-bromo-5-chlorobenzaldehyde
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: IIISHLMCTDMUHH-UHFFFAOYSA-N
SMILES: ClC1=CC(C=O)=C(Br)C=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)