missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorobenzaldehyde, 97%

Catalog No. AAH6433006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6433006 5 g
AAH6433014 25 g
AAH6433022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6433006 Supplier Thermo Scientific Chemicals Supplier No. H6433006

CAS: 174265-12-4 | C7H4BrClO | 219.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 174265-12-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00462870
InChI Key IIISHLMCTDMUHH-UHFFFAOYSA-N
Synonym 5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
PubChem CID 15391279
IUPAC Name 2-bromo-5-chlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)Br

Specifications

Melting Point 64°C to 68°C (literature)
Color Yellow
Quantity 5 g
Beilstein 10423891
Formula Weight 219.46
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Bromo-5-chlorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only