missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloroanisole 98.0+%, TCI America™

Catalog No. B36465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36465G 5 g
B364625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36465G Supplier TCI America Supplier No. B36465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174913-09-8
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.478
MDL Number MFCD03790889
InChI Key CQGYLDZGJLVLMK-UHFFFAOYSA-N
Synonym 2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzene
PubChem CID 17984845
IUPAC Name 1-bromo-4-chloro-2-methoxybenzene
SMILES COC1=C(C=CC(=C1)Cl)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 221.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-chloroanisole