missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
$96.26 - $475.52
Chemical Identifiers
| CAS | 328-89-2 |
|---|---|
| Molecular Formula | C8H4BrF3O2 |
| Molecular Weight (g/mol) | 269.017 |
| MDL Number | MFCD07368837 |
| InChI Key | SINIIFNWZPCJGU-UHFFFAOYSA-N |
| Synonym | 2-bromo-4-trifluoromethyl benzoic acid, 3-bromo-4-carboxybenzotrifluoride, benzoic acid, 2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl-benzoic acid, 2-bromo-4-trifluoromethylbenzoicacid, pubchem3184, acmc-209hwv, ksc495g8h, 2-bromo-4-trifluoromethyl-benzoic acid;2-brom-4-trifluormethyl-benzoesaeure;2-bromo-4-trifluoromethylbenzoic acid;2-brom-4-trifluormethylbenzolsaure; |
| PubChem CID | 67605 |
| IUPAC Name | 2-bromo-4-(trifluoromethyl)benzoic acid |
| SMILES | C1=CC(=C(C=C1C(F)(F)F)Br)C(=O)O |
Chemical Identifiers
| 328-89-2 | |
| 269.017 | |
| SINIIFNWZPCJGU-UHFFFAOYSA-N | |
| 67605 | |
| C1=CC(=C(C=C1C(F)(F)F)Br)C(=O)O |
| C8H4BrF3O2 | |
| MFCD07368837 | |
| 2-bromo-4-trifluoromethyl benzoic acid, 3-bromo-4-carboxybenzotrifluoride, benzoic acid, 2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl-benzoic acid, 2-bromo-4-trifluoromethylbenzoicacid, pubchem3184, acmc-209hwv, ksc495g8h, 2-bromo-4-trifluoromethyl-benzoic acid;2-brom-4-trifluormethyl-benzoesaeure;2-bromo-4-trifluoromethylbenzoic acid;2-brom-4-trifluormethylbenzolsaure; | |
| 2-bromo-4-(trifluoromethyl)benzoic acid |
Specifications
| 328-89-2 | |
| White-Yellow | |
| MFCD07368837 | |
| 2-bromo-4-trifluoromethyl benzoic acid, 3-bromo-4-carboxybenzotrifluoride, benzoic acid, 2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl-benzoic acid, 2-bromo-4-trifluoromethylbenzoicacid, pubchem3184, acmc-209hwv, ksc495g8h, 2-bromo-4-trifluoromethyl-benzoic acid;2-brom-4-trifluormethyl-benzoesaeure;2-bromo-4-trifluoromethylbenzoic acid;2-brom-4-trifluormethylbenzolsaure; | |
| C1=CC(=C(C=C1C(F)(F)F)Br)C(=O)O | |
| 269.017 | |
| 269.02 | |
| Crystalline Powder |
| 116°C | |
| C8H4BrF3O2 | |
| 1 g | |
| SINIIFNWZPCJGU-UHFFFAOYSA-N | |
| 2-bromo-4-(trifluoromethyl)benzoic acid | |
| 67605 | |
| ≥98.0% (GC,T) | |
| 2-Bromo-4-(trifluoromethyl)benzoic Acid |
Safety and Handling
TSCA : No