accessibility menu, dialog, popup

UserName

HOOBrFFF

CAS RN 328-89-2

IUPAC Name: 2-bromo-4-(trifluoromethyl)benzoic acid
Synonyms: 2-bromo-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 269.02
Molecular Formula: C8H4BrF3O2
InChi Key: SINIIFNWZPCJGU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1Br)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)