missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-(trifluoromethoxy)aniline 98.0+%, TCI America™
Supplier: TCI America B37995G
Specifications
| 2-Bromo-4-(trifluoromethoxy)aniline | |
| Yellow | |
| C7H5BrF3NO | |
| 5 g | |
| ROSTYHNIIDIBEG-UHFFFAOYSA-N | |
| 2-bromo-4-(trifluoromethoxy)aniline | |
| 688296 | |
| ≥98.0% (GC) |
| 175278-17-8 | |
| 209°C | |
| MFCD00203478 | |
| 2-bromo-4-trifluoromethoxy aniline, 2-bromo-4-tirfluoromethoxyaniline, 2-bromo-4-trifluoromethoxy-phenylamine, benzenamine, 2-bromo-4-trifluoromethoxy, 2-bromo-4-trifluoromethoxy phenylamine, acmc-1c8k7, ksc495e4t, 2-bromo-4 trifluoromethoxy aniline | |
| NC1=CC=C(OC(F)(F)F)C=C1Br | |
| 256.02 | |
| 256.02 | |
| Liquid |
Chemical Identifiers
| 175278-17-8 | |
| 256.02 | |
| ROSTYHNIIDIBEG-UHFFFAOYSA-N | |
| 688296 | |
| NC1=CC=C(OC(F)(F)F)C=C1Br |
| C7H5BrF3NO | |
| MFCD00203478 | |
| 2-bromo-4-trifluoromethoxy aniline, 2-bromo-4-tirfluoromethoxyaniline, 2-bromo-4-trifluoromethoxy-phenylamine, benzenamine, 2-bromo-4-trifluoromethoxy, 2-bromo-4-trifluoromethoxy phenylamine, acmc-1c8k7, ksc495e4t, 2-bromo-4 trifluoromethoxy aniline | |
| 2-bromo-4-(trifluoromethoxy)aniline |
Safety and Handling
TSCA : No