missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethoxy)aniline 98.0+%, TCI America™

Catalog No. p-7123173
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37995G 5 g
B379925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37995G Supplier TCI America Supplier No. B37995G

Chemical Identifiers

CAS 175278-17-8
Molecular Formula C7H5BrF3NO
Molecular Weight (g/mol) 256.02
MDL Number MFCD00203478
InChI Key ROSTYHNIIDIBEG-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethoxy aniline, 2-bromo-4-tirfluoromethoxyaniline, 2-bromo-4-trifluoromethoxy-phenylamine, benzenamine, 2-bromo-4-trifluoromethoxy, 2-bromo-4-trifluoromethoxy phenylamine, acmc-1c8k7, ksc495e4t, 2-bromo-4 trifluoromethoxy aniline
PubChem CID 688296
IUPAC Name 2-bromo-4-(trifluoromethoxy)aniline
SMILES NC1=CC=C(OC(F)(F)F)C=C1Br

Specifications

Color Yellow
Boiling Point 209°C
Quantity 5 g
Formula Weight 256.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-(trifluoromethoxy)aniline