missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7123711
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47371G 1 g
B47375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47371G Supplier TCI America Supplier No. B47371G
Only null left

Chemical Identifiers

CAS 66916-99-2
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD02664700
InChI Key XQELSBAAFMYSMG-UHFFFAOYSA-N
PubChem CID 3963668
IUPAC Name 2-(2-bromo-4-methoxyphenyl)acetic acid
SMILES COC1=CC(Br)=C(CC(O)=O)C=C1

Specifications

Melting Point 129°C
Color White
Quantity 1 g
UN Number 3077
Synonym 2-bromo-4-methoxyphenylacetic acid, 2-2-bromo-4-methoxyphenyl acetic acid, 2-bromo-4-methoxyphenyl acetic acid, benzeneacetic acid, 2-bromo-4-methoxy, 2-bromo-4-methoxyphenylacetci acid, pubchem3983, 2-2-bromo-4-methoxy-phenyl acetic acid, benzeneacetic acid,2-bromo-4-methoxy, 2-bromo-4-methoxy-phenyl-acetic acid
Formula Weight 245.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-methoxyphenylacetic Acid