accessibility menu, dialog, popup

UserName

CAS RN 66916-99-2

IUPAC Name: 2-(2-bromo-4-methoxyphenyl)acetic acid
Synonyms: (2-bromo-4-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: XQELSBAAFMYSMG-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(CC(O)=O)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)