missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxyphenylacetic acid, 97%

Catalog No. AAH2785903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2785903 1 g
AAH2785906 5 g
AAH2785914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2785903 Supplier Thermo Scientific Chemicals Supplier No. H2785903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66916-99-2 | C9H9BrO3 | 245.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66916-99-2
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD02664700
InChI Key XQELSBAAFMYSMG-UHFFFAOYSA-N
Synonym 2-bromo-4-methoxyphenylacetic acid, 2-2-bromo-4-methoxyphenyl acetic acid, 2-bromo-4-methoxyphenyl acetic acid, benzeneacetic acid, 2-bromo-4-methoxy, 2-bromo-4-methoxyphenylacetci acid, pubchem3983, 2-2-bromo-4-methoxy-phenyl acetic acid, benzeneacetic acid,2-bromo-4-methoxy, 2-bromo-4-methoxy-phenyl-acetic acid
PubChem CID 3963668
IUPAC Name 2-(2-bromo-4-methoxyphenyl)acetic acid
SMILES COC1=CC(Br)=C(CC(O)=O)C=C1

Specifications

Melting Point 127°C to 131°C
Quantity 1 g
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methoxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only