missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzylamine hydrochloride, 96%

Catalog No. AAH6220006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6220006 5 g
AAH6220003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6220006 Supplier Thermo Scientific Chemicals Supplier No. H6220006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 289038-14-8 | C7H8BrClFN | 240.5 g/mol

2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 289038-14-8
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.5
MDL Number MFCD00672922
InChI Key PBCSNHSXUSMNOV-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzylamine hydrochloride, 2-bromo-4-fluorophenyl methanamine hydrochloride, 1-2-bromo-4-fluorophenyl methanamine hydrochloride, 2-bromanyl-4-fluoranyl-phenyl methanamine hydrochloride
PubChem CID 2773353
IUPAC Name (2-bromo-4-fluorophenyl)methanamine;hydrochloride
SMILES C1=CC(=C(C=C1F)Br)CN.Cl

Specifications

Quantity 5 g
Formula Weight 240.5
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluorobenzylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only