CAS RN 289038-14-8
IUPAC Name:
1-(2-bromo-4-fluorophenyl)methanamine hydrochloride
Synonyms:
1-(2-bromo-4-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol):
240.5
Molecular Formula:
C7H8BrClFN
InChi Key:
PBCSNHSXUSMNOV-UHFFFAOYSA-N
SMILES:
Cl.NCC1=C(Br)C=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
2-Bromo-4-fluorobenzylamine hydrochloride, 96%
2-Bromo-4-fluorobenzylamine hydrochloride, 96%
CAS: 289038-14-8 Molecular Formula: C7H8BrClFN Molecular Weight (g/mol): 240.5 MDL Number: MFCD00672922 InChI Key: PBCSNHSXUSMNOV-UHFFFAOYSA-N Synonym: 2-bromo-4-fluorobenzylamine hydrochloride,2-bromo-4-fluorophenyl methanamine hydrochloride,1-2-bromo-4-fluorophenyl methanamine hydrochloride,2-bromanyl-4-fluoranyl-phenyl methanamine hydrochloride PubChem CID: 2773353 IUPAC Name: (2-bromo-4-fluorophenyl)methanamine;hydrochloride SMILES: C1=CC(=C(C=C1F)Br)CN.Cl