missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoroanisole, 98%

Catalog No. AAA1104009
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1104009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1104009 Supplier Thermo Scientific Chemicals Supplier No. A1104009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 452-08-4 | C7H6BrFO | 205.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 452-08-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD00012014
InChI Key JIQXVIJARQLCOY-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoroanisole, benzene, 2-bromo-4-fluoro-1-methoxy, 2-bromo-4-fluoro anisole, 1-bromo-3-fluoro-6-methoxybenzene, 2-bromo-4-fluoro-methoxybenzene, 2-bromo-4-fluoro-1-methoxy-benzene, 1-bromo-5-fluoro-2-methoxybenzene, attercop-chm at106665, pubchem1972, 2-bromo-4-fluoroanisol
PubChem CID 136292
IUPAC Name 2-bromo-4-fluoro-1-methoxybenzene
SMILES COC1=CC=C(F)C=C1Br

Specifications

Density 1.581
Boiling Point 88°C to 89°C (1 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.545
Quantity 10 g
Beilstein 2358880
Formula Weight 205.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoroanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only