accessibility menu, dialog, popup

UserName

H3COFBr

CAS RN 452-08-4

IUPAC Name: 2-bromo-4-fluoro-1-methoxybenzene
Synonyms: 2-bromo-4-fluoro-1-methoxybenzene
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: JIQXVIJARQLCOY-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C=C1Br
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)