missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-1-nitrobenzene 98.0+%, TCI America™

Catalog No. B30615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30615G Supplier TCI America Supplier No. B30615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 700-36-7
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD00792441
InChI Key VGYVBEJDXIPSDL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoronitrobenzene, 1-bromo-5-fluoro-2-nitrobenzene, 3-bromo-1-fluoro-4-nitrobenzene, 4-fluoro-2-bromonitrobenzene, benzene, 2-bromo-4-fluoro-1-nitro, 4-fluoro-2-bromo nitrobenzene, 2-bromo-4-fluoro-1-nitro-benzene, zlchem 479, pubchem4277, acmc-1bkfy
PubChem CID 2756993
IUPAC Name 2-bromo-4-fluoro-1-nitrobenzene
SMILES C1=CC(=C(C=C1F)Br)[N+](=O)[O-]

Specifications

Melting Point 44°C
Quantity 5 g
Formula Weight 220.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluoro-1-nitrobenzene