accessibility menu, dialog, popup

UserName

FBrNOO

CAS RN 700-36-7

IUPAC Name: 2-bromo-4-fluoro-1-nitrobenzene
Synonyms: 2-bromo-4-fluoro-1-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: VGYVBEJDXIPSDL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Br)C=C(F)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)