missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-1-nitrobenzene, 98%

Catalog No. p-7053787
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6146606 5 g
AAH6146614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6146606 Supplier Thermo Scientific Chemicals Supplier No. H6146606
Only null left

CAS: 700-36-7 | C6H3BrFNO2 | 219.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-36-7
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD00792441
InChI Key VGYVBEJDXIPSDL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoronitrobenzene, 1-bromo-5-fluoro-2-nitrobenzene, 3-bromo-1-fluoro-4-nitrobenzene, 4-fluoro-2-bromonitrobenzene, benzene, 2-bromo-4-fluoro-1-nitro, 4-fluoro-2-bromo nitrobenzene, 2-bromo-4-fluoro-1-nitro-benzene, zlchem 479, pubchem4277, acmc-1bkfy
PubChem CID 2756993
IUPAC Name 2-bromo-4-fluoro-1-nitrobenzene
SMILES C1=CC(=C(C=C1F)Br)[N+](=O)[O-]

Specifications

Melting Point 41°C to 45°C
Color Green to Yellow
Quantity 5 g
Formula Weight 220
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluoro-1-nitrobenzene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only