missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-4-chlorobiphenyl 98.0+%, TCI America™

Catalog No. B52531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52531G Supplier TCI America Supplier No. B52531G
Only null left

Chemical Identifiers

CAS 179526-95-5
Molecular Formula C12H8BrCl
Molecular Weight (g/mol) 267.55
MDL Number MFCD18072809
InChI Key WSHZWUXRWQVZQP-UHFFFAOYSA-N
PubChem CID 12939405
IUPAC Name 2-bromo-4'-chloro-1,1'-biphenyl
SMILES ClC1=CC=C(C=C1)C1=CC=CC=C1Br

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 1 g
UN Number 3152
Formula Weight 267.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2′-Bromo-4-chlorobiphenyl